Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ms4a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514749.1 | XP_003514797.1 | 1 - 2 | 43 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |