Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mad2l1bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003515163.2 | XP_003515211.1 | 1 - 2 | 131 | 75 | |
GenBank mRNA | JI879553.1 | - | 1 - 2 | 95 | 75 | |
JP064344.1 | - | 1 - 2 | 95 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Probe spans exons |