Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sage1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007640578.1 | XP_007638768.1 | 5 - 6 | 805 | 94 | |
GenBank mRNA | JI893420.1 | - | 1 - 2 | 104 | 94 | |
JP044505.1 | - | 1 - 2 | 104 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |