Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3galt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003513238.2 | XP_003513286.2 | 2 | 894 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Both primers and probe map within a single exon |