Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496017.2 | XP_003496065.1 | 3 - 4 | 567 | 98 | |
XM_007646261.1 | XP_007644451.1 | 3 - 4 | 567 | 98 | ||
XM_007646270.1 | XP_007644460.1 | 3 - 4 | 489 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Protease 3' Most Probe spans exons |