Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pygo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501598.2 | XP_003501646.1 | 1 - 2 | 112 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |