Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502053.2 | XP_003502101.1 | 2 - 3 | 189 | 78 | |
GenBank mRNA | JP060519.1 | - | 2 - 3 | 179 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |