Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Odf3l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502552.1 | XP_003502600.1 | 3 - 4 | 349 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |