Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh3b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007650570.1 | XP_007648760.1 | 7 - 8 | 1038 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Probe spans exons |