Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007640388.1 | XP_007638578.1 | 2 - 3 | 408 | 92 | |
GenBank mRNA | JI866009.1 | - | 2 - 3 | 218 | 92 |