Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007640388.1 | XP_007638578.1 | 3 - 4 | 824 | 67 | |
GenBank mRNA | JI866009.1 | - | 3 - 4 | 634 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |