Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Znrf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507796.2 | XP_003507844.1 | 2 - 3 | 160 | 99 | |
GenBank mRNA | JP053232.1 | - | 1 - 2 | 105 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |