Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007634346.1 | XP_007632536.1 | 2 - 3 | 272 | 76 | |
GenBank mRNA | JP057234.1 | - | 2 - 3 | 272 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |