Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003513871.2 | XP_003513919.1 | 4 - 5 | 1482 | 89 | |
XM_007654102.1 | XP_007652292.1 | 4 - 5 | 1482 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Protease Probe spans exons |