Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007647718.1 | XP_007645908.1 | 2 - 3 | 310 | 100 | |
GenBank mRNA | JP043458.1 | - | 2 - 3 | 203 | 100 |