Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dyrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497144.2 | XP_003497192.2 | 1 - 2 | 41 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Protein Kinase 3' Most Probe spans exons |