Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ralb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499436.2 | XP_003499484.1 | 2 - 3 | 276 | 103 | |
XM_007640207.1 | XP_007638397.1 | 2 - 3 | 315 | 103 | ||
GenBank mRNA | JP044159.1 | - | 2 - 3 | 225 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |