Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508554.2 | XP_003508602.1 | 8 - 9 | 823 | 66 | |
XM_007649181.1 | XP_007647371.1 | 8 - 9 | 823 | 66 | ||
GenBank mRNA | JP056583.1 | - | 7 - 8 | 645 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |