Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahsp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514786.2 | XP_003514834.1 | 2 - 3 | 251 | 104 | |
GenBank mRNA | JI886446.1 | - | 3 - 4 | 340 | 104 | |
JP040857.1 | - | 3 - 4 | 340 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |