Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcna4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497560.1 | XP_003497608.1 | 1 | 1714 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Ion Channel 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |