Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snai3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495092.1 | XP_003495140.1 | 2 - 3 | 684 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |