Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498294.2 | XP_003498342.1 | 2 - 3 | 240 | 83 | |
XM_007625783.1 | XP_007623973.1 | 2 - 3 | 249 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |