Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fibcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003513748.1 | XP_003513796.1 | 1 - 2 | 48 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |