Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tomm20l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003515625.2 | XP_003515673.1 | 1 - 2 | 141 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |