Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007611633.1 | XP_007609823.1 | 12 - 13 | 3264 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Probe spans exons |