Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508770.2 | XP_003508818.1 | 3 - 4 | 275 | 86 | |
XM_007649394.1 | XP_007647584.1 | 3 - 4 | 238 | 86 | ||
GenBank mRNA | JI871534.1 | - | 1 - 2 | 73 | 86 | |
JP065150.1 | - | 1 - 2 | 73 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |