Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007609980.1 | XP_007608170.1 | 3 - 4 | 1014 | 69 | |
GenBank mRNA | JP048266.1 | - | 1 - 2 | 206 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Probe spans exons |