Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map1s | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007638529.1 | XP_007636719.1 | 5 - 6 | 2663 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |