Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map1s | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007638529.1 | XP_007636719.1 | 6 - 7 | 2886 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |