Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643211.1 | XP_007641401.1 | 3 - 4 | 332 | 55 | |
XM_007643212.1 | XP_007641402.1 | 3 - 4 | 1059 | 55 | ||
XM_007643213.1 | XP_007641403.1 | 4 - 5 | 569 | 55 | ||
GenBank mRNA | JI883595.1 | - | 3 - 4 | 373 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons |