Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496159.2 | XP_003496207.1 | 2 - 3 | 519 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Probe spans exons |