Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496522.1 | XP_003496570.1 | 3 - 4 | 980 | 126 | |
XM_007650933.1 | XP_007649123.1 | 3 - 4 | 977 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |