Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vgll1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499823.1 | XP_003499871.1 | 1 - 2 | 217 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |