Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Angptl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496150.2 | XP_003496198.1 | 7 - 8 | 1021 | 124 | |
XM_007647313.1 | XP_007645503.1 | 5 - 6 | 826 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |