Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007653259.1 | XP_007651449.1 | 1 - 2 | 218 | 76 | |
GenBank mRNA | JI892498.1 | - | 1 | 218 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |