Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx39a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501883.2 | XP_003501931.1 | 10 - 11 | 1371 | 111 | |
GenBank mRNA | JI892008.1 | - | 4 - 5 | 537 | 111 | |
JP055369.1 | - | 5 - 6 | 598 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Probe spans exons |