Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx39a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501883.2 | XP_003501931.1 | 5 - 6 | 722 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |