Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501889.2 | XP_003501937.1 | 2 - 3 | 376 | 69 | |
GenBank mRNA | JI873565.1 | - | 2 - 3 | 375 | 69 | |
JP041437.1 | - | 2 - 3 | 375 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |