Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pard6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508555.1 | XP_003508603.1 | 1 - 2 | 65 | 70 | |
XM_007649183.1 | XP_007647373.1 | 1 - 2 | 65 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |