Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rangrf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497172.2 | XP_003497220.1 | 3 - 4 | 734 | 78 | |
XM_007608217.1 | XP_007606407.1 | 2 - 3 | 829 | 78 | ||
XM_007608221.1 | XP_007606411.1 | 2 - 3 | 827 | 78 | ||
XM_007608224.1 | XP_007606414.1 | 2 - 3 | 835 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |