Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497386.2 | XP_003497434.1 | 2 - 3 | 222 | 85 | |
GenBank mRNA | JI871269.1 | - | 2 - 3 | 190 | 85 | |
JP065117.1 | - | 2 - 3 | 190 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |