Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Magt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003505297.2 | XP_003505345.1 | 6 - 7 | 809 | 73 | |
GenBank mRNA | JI885607.1 | - | 6 - 7 | 809 | 73 | |
JP055298.1 | - | 6 - 7 | 809 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |