Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tctex1d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495399.2 | XP_003495447.1 | 2 - 3 | 358 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |