Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ckmt1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498034.2 | XP_003498082.1 | 4 - 5 | 867 | 63 | |
XM_003498035.2 | XP_003498083.1 | 5 - 6 | 809 | 63 | ||
XM_007621602.1 | XP_007619792.1 | 5 - 6 | 861 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |