Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ooep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503187.1 | XP_003503235.1 | 2 - 3 | 371 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |