Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003512152.2 | XP_003512200.1 | 1 | 2321 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Both primers and probe map within a single exon |