Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499538.2 | XP_003499586.1 | 4 - 5 | 880 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |