Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Avpr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500787.1 | XP_003500835.1 | 1 - 2 | 978 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Recommended Assay Probe spans exons |