Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tenc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007655053.1 | XP_007653243.1 | 13 - 14 | 1773 | 80 | |
GenBank mRNA | JI887784.1 | - | 2 - 3 | 221 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |