Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tenc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007655053.1 | XP_007653243.1 | 14 - 15 | 1838 | 83 | |
GenBank mRNA | JI887784.1 | - | 3 - 4 | 286 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |